1-(2-Morpholinoethyl)-1H-pyrazol-4-boronsäurepinakolester 98%
Lieferant: Apollo Scientific
Synonyme:
4-(2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)ethyl)morpholine
OR17572-1G
OR17572-5G
APOSOR17572-1GEA
112
EUR
APOSOR17572-1G
APOSOR17572-5G
1-(2-Morpholinoethyl)-1H-pyrazol-4-boronsäurepinakolester 98%
1-(2-Morpholinoethyl)-1H-pyrazol-4-boronsäurepinakolester
Formel:
C₁₅H₂₆BN₃O₃ Molecular Weight: 307.1962 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08062342 CAS-Nummer: 864754-18-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...