3-Methyl-1-benzofuran-2-carbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR17626-100G
OR17626-25G
APOSOR17626-100GEA
282
EUR
APOSOR17626-100G
APOSOR17626-25G
3-Methyl-1-benzofuran-2-carbonsäure
3-Methyl-1-benzofuran-2-carbonsäure
Formel:
C₁₀H₈O₃ Schmelzpunkt: 194…197 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060511 CAS-Nummer: 24673-56-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...