1H-Indazole-4-carbaldehyde 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Formylindazole, 1H-Indazole-4-carboxaldehyde, 4-Formyl-1H-indazole

OR17651-1G OR17651-5G
APOSOR17651-1GEA 55.1 EUR
APOSOR17651-1G APOSOR17651-5G
1H-Indazole-4-carbaldehyde 95%
1H-Indazole-4-carbaldehyde
Formel: C₈H₆N₂O
Molecular Weight: 146,15 g/mol
Schmelzpunkt: 166…167 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06738279
CAS-Nummer: 669050-70-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR