5-Cyano-3-pyridinylboronic acid 98%

Lieferant: Apollo Scientific
Achtung

OR17709-1G OR17709-250MG OR17709-5G
APOSOR17709-1GEA 55.1 EUR
APOSOR17709-1G APOSOR17709-250MG APOSOR17709-5G
5-Cyano-3-pyridinylboronic acid 98%
5-Cyano-3-pyridinylboronic acid
Formel: C6H5BN2O2
Molecular Weight: 147,9300 g/mol
Schmelzpunkt: 143…145 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07374882
CAS-Nummer: 497147-93-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR