3-Brom-5-methoxybenzoesäure 99%
Lieferant: Apollo Scientific
|
Achtung
|
OR17818-1G
OR17818-25G
OR17818-5G
APOSOR17818-1GEA
21.9
EUR
APOSOR17818-1G
APOSOR17818-25G
APOSOR17818-5G
3-Brom-5-methoxybenzoesäure 99%
3-Bromo-5-methoxybenzoic acid
Formel:
C₈H₇BrO₃ Schmelzpunkt: 192…193 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00266858 CAS-Nummer: 157893-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...