2,4-Dichlor-6-iodanilin
Lieferant: Apollo Scientific
|
Achtung
|
OR17819-100G
OR17819-25G
OR17819-5G
APOSOR17819-100GEA
178
EUR
APOSOR17819-100G
APOSOR17819-25G
APOSOR17819-5G
2,4-Dichlor-6-iodanilin
2,4-Dichlor-6-iodanilin
Formel:
C₆H₄Cl₂IN Molecular Weight: 287,92 g/mol Schmelzpunkt: 79…81 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08533279 CAS-Nummer: 697-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...