4-(4-Bromphenyl)morpholin

Lieferant: Apollo Scientific
Achtung

OR17823-100G OR17823-25G
APOSOR17823-100GEA 267 EUR
APOSOR17823-100G APOSOR17823-25G
4-(4-Bromphenyl)morpholin
4-(4-Bromphenyl)morpholin
Formel: C₁₀H₁₂BrNO
Molecular Weight: 242.12 g/mol
Schmelzpunkt: 114…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04112483
CAS-Nummer: 30483-75-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR