Aniline-2,5-disulphonic acid monosodium salt
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Sodium Hydrogen 2-Amino-1,4-benzenedisulphonate, 2,5-Disulfoaniline Monosodium Salt, Aniline-2,5-disulphonic Acid Monosodium Salt, 2,5-Disulphoaniline Monosodium Salt, Sodium Hydrogen 2-Amino-1,4-benzenedisulfonate, 2-Amino-1,4-benzenedisulfonic acid Monosodium Salt, Sodium 2-amino-4-sulphobenzenesulphonate, 2-Amino-1,4-benzenedisulphonic acid Monosodium Salt
OR17827-25G
OR17827-5G
OR17827-100G
APOSOR17827-25GEA
71.2
EUR
APOSOR17827-25G
APOSOR17827-5G
APOSOR17827-100G
Aniline-2,5-disulphonic acid monosodium salt
Aniline-2,5-disulphonic acid monosodium salt
Formel:
C₆H₇NO₆S₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078393 CAS-Nummer: 24605-36-5 EINECS: 246-349-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...