2,5-Dibrom-3-hexylthiophen 98%

Lieferant: Apollo Scientific

OR17832-25G OR17832-1G OR17832-5G
APOSOR17832-25GEA 231 EUR
APOSOR17832-25G APOSOR17832-1G APOSOR17832-5G
2,5-Dibrom-3-hexylthiophen 98%
2,5-Dibromo-3-hexylthiophene
Formel: C₁₀H₁₄Br₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00274313
CAS-Nummer: 116971-11-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR