5-Aminomethyl-2-(boc-amino)pyridine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-5-(aminomethyl)pyridine, 2-BOC protected, 2-(Boc-amino)-5-(aminomethyl)pyridine

OR17840-1G OR17840-250MG
APOSOR17840-1GEA 227 EUR
APOSOR17840-1G APOSOR17840-250MG
5-Aminomethyl-2-(boc-amino)pyridine 95%
5-Aminomethyl-2-(boc-amino)pyridine
Formel: C₁₁H₁₇N₃O₂
Molecular Weight: 223,28 g/mol
Schmelzpunkt: 141…143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07781132
CAS-Nummer: 187237-37-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR