2-(Brommethyl)chinolin

Lieferant: Apollo Scientific
Gefahr

OR17841-1G OR17841-250MG
APOSOR17841-1GEA 55.1 EUR
APOSOR17841-1G APOSOR17841-250MG
2-(Brommethyl)chinolin
2-(Bromomethyl)quinoline
Formel: C₁₀H₈BrN
Molecular Weight: 222.08 g/mol
Schmelzpunkt: 55…56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08062399
CAS-Nummer: 5632-15-5
UN: 3077
ADR: 9+EHS III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR