2-Methyl-2H-pyrazole-3-sulphonyl chloride 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-Methyl-1H-pyrazole-5-sulphonyl chloride

OR17845-1G OR17845-250MG
APOSOR17845-1GEA 230 EUR
APOSOR17845-1G APOSOR17845-250MG
2-Methyl-2H-pyrazole-3-sulphonyl chloride 95%
2-Methyl-2H-pyrazole-3-sulphonyl chloride
Formel: C₄H₅ClN₂O₂S
Molecular Weight: 180.61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09065021
CAS-Nummer: 1020721-61-2
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR