3-Iod-p-toluylsäure

Lieferant: Apollo Scientific
Achtung

OR17850-100G OR17850-25G OR17850-5G
APOSOR17850-100GEA 82.8 EUR
APOSOR17850-100G APOSOR17850-25G APOSOR17850-5G
3-Iod-p-toluylsäure
3-Iod-p-toluylsäure
Formel: C₈H₇IO₂
Molecular Weight: 262,05 g/mol
Schmelzpunkt: 208…212 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010392
CAS-Nummer: 82998-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR