5-Amino-2-chlorbenzonitril
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chloro-3-cyanoaniline
OR17860-25G
APOSOR17860-25GEA
159
EUR
APOSOR17860-25G
5-Amino-2-chlorbenzonitril
5-Amino-2-chlorbenzonitril
Formel:
C₇H₅ClN₂ Molecular Weight: 152,5800 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00800459 CAS-Nummer: 35747-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...