5-(Hydroxymethyl)pyrimidine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Pyrimidinylmethanol
OR17898-250MG
OR17898-1G
OR17898-5G
APOSOR17898-250MGEA
26.1
EUR
APOSOR17898-250MG
APOSOR17898-1G
APOSOR17898-5G
5-(Hydroxymethyl)pyrimidine 97%
5-(Hydroxymethyl)pyrimidine
Formel:
C₅H₆N₂O Molecular Weight: 110,1100 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD03789623 CAS-Nummer: 25193-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...