2-(4-Hydroxyphenyl)propionsäure

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-(4-Hydroxyphenyl)propionic Acid

OR17909-10G OR17909-1G OR17909-50G
APOSOR17909-10GEA 121 EUR
APOSOR17909-10G APOSOR17909-1G APOSOR17909-50G
2-(4-Hydroxyphenyl)propionsäure
2-(4-Hydroxyphenyl)propionsäure
Formel: C₉H₁₀O₃
Molecular Weight: 166.18 g/mol
Schmelzpunkt: 130…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00128847
CAS-Nummer: 938-96-5
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR