4-Chinolinamin 95+%

Lieferant: Apollo Scientific
Achtung

OR17934-1G OR17934-5G OR17934-10G
APOSOR17934-1GEA 29 EUR
APOSOR17934-1G APOSOR17934-5G APOSOR17934-10G
4-Chinolinamin 95+%
4-Aminoquinoline
Formel: C₉H₈N₂
Molecular Weight: 144,1700 g/mol
Schmelzpunkt: 152…154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00463448
CAS-Nummer: 578-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR