4-Brom-1-iod-2-nitrobenzol 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2-iodonitrobenzene

OR17993-25G OR17993-5G OR17993-100G
APOSOR17993-25GEA 48 EUR
APOSOR17993-25G APOSOR17993-5G APOSOR17993-100G
4-Brom-1-iod-2-nitrobenzol 99%
4-Brom-1-iod-2-nitrobenzol
Formel: C₆H₃BrINO₂
Molecular Weight: 327,9020 g/mol
Schmelzpunkt: 87…89 °C
Lagertemperatur: Kühlschrank
MDL: MFCD11505492
CAS-Nummer: 112671-42-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR