8-Amino-4-methylchinolin

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Methylquinolin-8-amine

OR18011-250MG OR18011-500MG
APOSOR18011-250MGEA 120 EUR
APOSOR18011-250MG APOSOR18011-500MG
8-Amino-4-methylchinolin
8-Amino-4-methylquinoline
Formel: C₁₀H₁₀N₂
Lagertemperatur: Kühlschrank
MDL: MFCD03819702
CAS-Nummer: 62748-01-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR