3-Brom-2-thiophencarbonitril 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Bromothiophene-2-carbonitrile
OR18012-1G
OR18012-5G
OR18012-250MG
APOSOR18012-1GEA
37.7
EUR
APOSOR18012-1G
APOSOR18012-5G
APOSOR18012-250MG
3-Brom-2-thiophencarbonitril 95%
3-Brom-2-thiophencarbonitril
Formel:
C₅H₂BrNS Molecular Weight: 188,05 g/mol Schmelzpunkt: 43…48 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00068173 CAS-Nummer: 18791-98-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...