Methyl-3-amino-2-hydroxy-5-methylbenzenecarboxylate 95+%

Lieferant: Apollo Scientific
Achtung

OR18028-1G OR18028-500MG OR18028-5G
APOSOR18028-1GEA 336 EUR
APOSOR18028-1G APOSOR18028-500MG APOSOR18028-5G
Methyl-3-amino-2-hydroxy-5-methylbenzenecarboxylate 95+%
Methyl-3-amino-2-hydroxy-5-methylbenzenecarboxylate
Formel: C₉H₁₁NO₃
Molecular Weight: 181,19 g/mol
Schmelzpunkt: 72…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD14560550
CAS-Nummer: 70978-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR