5-Bromo-2-chloro-6-cyclopropylnicotinonitrile

Lieferant: Apollo Scientific
Achtung

OR18050-1G OR18050-500MG
APOSOR18050-1GEA 920 EUR
APOSOR18050-1G APOSOR18050-500MG
5-Bromo-2-chloro-6-cyclopropylnicotinonitrile
5-Bromo-2-chloro-6-cyclopropylnicotinonitrile
Formel: C₉H₆BrClN₂
Molecular Weight: 257.52 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14584776
CAS-Nummer: 1221792-18-2
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR