7-Brom-1-tetralon 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Bromo-3,4-dihydronaphthalen-1(2H)-one
OR18054-100G
OR18054-25G
OR18054-5G
APOSOR18054-100GEA
219
EUR
APOSOR18054-100G
APOSOR18054-25G
APOSOR18054-5G
7-Brom-1-tetralon 99%
7-Brom-1-tetralon
Formel:
C₁₀H₉BrO Molecular Weight: 225,08 g/mol Schmelzpunkt: 75…80 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02179287 CAS-Nummer: 32281-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...