2-Bromo-5-cyanopyrazine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromopyrazine-2-carbonitrile

OR18105-1G OR18105-250MG
APOSOR18105-1GEA 33.5 EUR
APOSOR18105-1G APOSOR18105-250MG
2-Bromo-5-cyanopyrazine
2-Bromo-5-cyanopyrazine
Formel: C₅H₂BrN₃
Molecular Weight: 184 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09909652
CAS-Nummer: 221295-04-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR