Ethyl-1H-1,3-benzimidazole-2-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

OR18118-1G OR18118-5G
APOSOR18118-1GEA 46.4 EUR
APOSOR18118-1G APOSOR18118-5G
Ethyl-1H-1,3-benzimidazole-2-carboxylate 95%
Ethyl-1H-1,3-benzimidazole-2-carboxylate
Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190,2 g/mol
Schmelzpunkt: 185…188 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00721392
CAS-Nummer: 1865-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR