Methyl-2-brom-4-nitrobenzoat 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Brom-4-nitrobenzoesäuremethylester
OR18260-1G
OR18260-25G
OR18260-5G
APOSOR18260-1GEA
21.9
EUR
APOSOR18260-1G
APOSOR18260-25G
APOSOR18260-5G
Methyl-2-brom-4-nitrobenzoat 97%
Methyl-2-brom-4-nitrobenzoat
Formel:
C₈H₆BrNO₄ Molecular Weight: 260,04 g/mol Schmelzpunkt: 84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07779271 CAS-Nummer: 100959-22-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...