4,6-Dichloro-2-methoxypyrimidine 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR18261-100MG
OR18261-1G
APOSOR18261-100MGEA
21.9
EUR
APOSOR18261-100MG
APOSOR18261-1G
4,6-Dichloro-2-methoxypyrimidine 98%
4,6-Dichloro-2-methoxypyrimidine
Formel:
C₅H₄Cl₂N₂O Molecular Weight: 179 g/mol Schmelzpunkt: 53…55 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD03428360 CAS-Nummer: 1074-40-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...