1,1'-Binaphthalin-2,2'-diylbis(diphenylphosphin) 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: racemic-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, rac-BINAP, (+/-)-(1,1'-Binaphthalene-2,2'-diyl)bis(diphenylphosphine), (±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (+/-)-BINAP

OR18264-25G OR18264-5G
APOSOR18264-25GEA 66.7 EUR
APOSOR18264-25G APOSOR18264-5G
1,1'-Binaphthalin-2,2'-diylbis(diphenylphosphin) 98%
2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Formel: C₄₄H₃₂P₂
Molecular Weight: 622,67 g/mol
Schmelzpunkt: 283…286 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010805
CAS-Nummer: 98327-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR