(2-Amino-5-chlorophenyl)phenylmethanol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-5-chlorobenzhydrol

OR18268-1G OR18268-250MG
APOSOR18268-1GEA 464 EUR
APOSOR18268-1G APOSOR18268-250MG
(2-Amino-5-chlorophenyl)phenylmethanol
(2-Amino-5-chlorophenyl)phenylmethanol
Formel: C₁₃H₁₂ClNO
Molecular Weight: 233,7 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01004640
CAS-Nummer: 7039-50-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR