Sodium-1-naphthalenesulphonate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Naphthalenesulphonic acid sodium salt, Naphthalene-1-sulphonic acid sodium salt

OR18282-100G OR18282-25G OR18282-500G
APOSOR18282-100GEA 43.5 EUR
APOSOR18282-100G APOSOR18282-25G APOSOR18282-500G
Sodium-1-naphthalenesulphonate 98%
Sodium-1-naphthalenesulphonate
Formel: C₁₀H₈O₃S
Molecular Weight: 230.21 g/mol
Schmelzpunkt: 299…301 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064964
CAS-Nummer: 130-14-3
EINECS: 204-976-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR