4,4'-Bis(diethylamino)benzophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Michlers Ethylketon, 4,4'-Di(diethylamino)benzophenon
OR18283-100G
OR18283-25G
OR18283-500G
APOSOR18283-100GEA
39.3
EUR
APOSOR18283-100G
APOSOR18283-25G
APOSOR18283-500G
4,4'-Bis(diethylamino)benzophenon 98%
4,4'-Bis(diethylamino)benzophenon
Formel:
C₂₁H₂₈N₂O Molecular Weight: 324,47 g/mol Schmelzpunkt: 89…95 °C Flammpunkt: 151 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009044 CAS-Nummer: 90-93-7 EINECS: 202-025-4 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...