(4-Pyridylthio)essigsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Pyridin-4-ylthio)acetic acid
OR18288-100G
OR18288-500G
APOSOR18288-100GEA
46.4
EUR
APOSOR18288-100G
APOSOR18288-500G
(4-Pyridylthio)essigsäure 98%
(4-Pyridylthio)essigsäure
Formel:
C₇H₇NO₂S Molecular Weight: 169,2 g/mol Schmelzpunkt: 251…260 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006424 CAS-Nummer: 10351-19-6 EINECS: 233-765-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...