1-(Hydroxymethyl)-5,5-dimethyl-2,4-imidazolidindion 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5,5-Dimethyl-1-(hydroxymethyl)hydantoin

OR18297-100G OR18297-25G
APOSOR18297-100GEA 24.8 EUR
APOSOR18297-100G APOSOR18297-25G
1-(Hydroxymethyl)-5,5-dimethyl-2,4-imidazolidindion 97%
1-(Hydroxymethyl)-5,5-dimethylhydantoin
Formel: C₆H₁₀N₂O₃
Molecular Weight: 158,16 g/mol
Schmelzpunkt: 104…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00020866
CAS-Nummer: 116-25-6
EINECS: 204-132-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR