6-Chloroxindol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Chloro-2-oxindole
OR18299-25G
OR18299-5G
APOSOR18299-25GEA
24.8
EUR
APOSOR18299-25G
APOSOR18299-5G
6-Chloroxindol 97%
6-Chloroxindol
Formel:
C₈H₆ClNO Molecular Weight: 167,59 g/mol Schmelzpunkt: 195…199 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00209962 CAS-Nummer: 56341-37-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...