2-Aminobenzothiazol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzothiazol-2-ylamin, Cer(III)-Ionophor II

OR18345-100G OR18345-25G
APOSOR18345-100GEA 36.4 EUR
APOSOR18345-100G APOSOR18345-25G
2-Aminobenzothiazol 97%
2-Aminobenzothiazole
Formel: C₇H₆N₂S
Molecular Weight: 150,2 g/mol
Schmelzpunkt: 126…129 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005785
CAS-Nummer: 136-95-8
EINECS: 205-268-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR