2-Nitro-5-piperidinoaniline
Lieferant: Apollo Scientific
OR183519-1G
OR183519-5G
APOSOR183519-1GEA
85.7
EUR
APOSOR183519-1G
APOSOR183519-5G
2-Nitro-5-piperidinoaniline
2-Nitro-5-piperidinoaniline
Formel:
C₁₁H₁₅N₃O₂ Molecular Weight: 221,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00545948 CAS-Nummer: 54997-99-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...