2,4-Dichlorbenzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
OR18360-1KG
OR18360-100G
OR18360-500G
APOSOR18360-1KGEA
66.7
EUR
APOSOR18360-1KG
APOSOR18360-100G
APOSOR18360-500G
2,4-Dichlorbenzoesäure
2,4-Dichlorobenzoic acid
Formel:
C₇H₄Cl₂O₂ Molecular Weight: 191,01 g/mol Schmelzpunkt: 159…165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002414 CAS-Nummer: 50-84-0 EINECS: 200-067-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...