Tritylchlorid 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Triphenylmethylchlorid, Triphenylchlormethan, Chlortriphenylmethan
OR18363-100G
OR18363-500G
OR18363-1KG
APOSOR18363-100GEA
21.9
EUR
APOSOR18363-100G
APOSOR18363-500G
APOSOR18363-1KG
Tritylchlorid 95%
Tritylchlorid
Formel:
C₁₉H₁₅Cl Molecular Weight: 278,78 g/mol Siedepunkt: 230…235 °C/20mmHg Schmelzpunkt: 111…113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000813 CAS-Nummer: 76-83-5 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...