3,5-Dihydroxybenzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
alpha-Resorcylic acid
OR18364-100G
OR18364-500G
OR18364-25G
APOSOR18364-100GEA
65.4
EUR
APOSOR18364-100G
APOSOR18364-500G
APOSOR18364-25G
3,5-Dihydroxybenzoesäure 98%
3,5-Dihydroxybenzoic acid
Formel:
C₇H₆O₄ Molecular Weight: 154,12 g/mol Schmelzpunkt: 236…238 °C Flammpunkt: 200 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002512 CAS-Nummer: 99-10-5 EINECS: 202-730-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...