3-Acetylbenzonitril 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl-m-cyanphenylketon, 3'-Cyanacetophenon

OR18369-25G
APOSOR18369-25GEA 72.5 EUR
APOSOR18369-25G
3-Acetylbenzonitril 98%
3-Acetylbenzonitril
Formel: C₉H₇NO
Molecular Weight: 145,16 g/mol
Schmelzpunkt: 97…101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001806
CAS-Nummer: 6136-68-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR