4-(Diethylamino)benzoesäure 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Diethylaminobenzoic Acid

OR18377-5G OR18377-1G OR18377-25G
APOSOR18377-5GEA 24.8 EUR
APOSOR18377-5G APOSOR18377-1G APOSOR18377-25G
4-(Diethylamino)benzoesäure 99%
4-(Diethylamino)benzoesäure
Formel: C₁₁H₁₅NO₂
Molecular Weight: 193,25 g/mol
Schmelzpunkt: 192…195 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002538
CAS-Nummer: 5429-28-7
EINECS: 226-582-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR