L(-)-Carnitin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate

OR18381-100G OR18381-25G OR18381-500G
APOSOR18381-100GEA 36.4 EUR
APOSOR18381-100G APOSOR18381-25G APOSOR18381-500G
L(-)-Carnitin 98%
L(-)-Carnitin
Formel: C₇H₁₅NO₃
Molecular Weight: 161,20 g/mol
Schmelzpunkt: 210…212 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00038747
CAS-Nummer: 541-15-1
EINECS: 208-768-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR