3-Iod-4-methyl-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
OR18424-1G
OR18424-5G
APOSOR18424-1GEA
75.4
EUR
APOSOR18424-1G
APOSOR18424-5G
3-Iod-4-methyl-1H-indazol
3-Iodo-4-methyl-1H-indazole
Formel:
C₈H₇IN₂ Molecular Weight: 258,0591 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07781537 CAS-Nummer: 885522-63-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...