3-Iod-5-methyl-1H-indazol

Lieferant: Apollo Scientific
Achtung

OR18425-1G OR18425-250MG
APOSOR18425-1GEA 62.5 EUR
APOSOR18425-1G APOSOR18425-250MG
3-Iod-5-methyl-1H-indazol
3-Iodo-5-methyl-1H-indazole
Formel: C₈H₇IN₂
Molecular Weight: 258,0600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781621
CAS-Nummer: 885518-92-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR