Methyl 2-Nitrophenyl sulphone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(Methylsulphonyl)-2-nitrobenzene
OR18540-1G
OR18540-5G
OR18540-10G
APOSOR18540-1GEA
27.7
EUR
APOSOR18540-1G
APOSOR18540-5G
APOSOR18540-10G
Methyl 2-Nitrophenyl sulphone
Methyl 2-Nitrophenyl sulphone
Formel:
C₇H₇NO₄S Molecular Weight: 201,20 g/mol Schmelzpunkt: 102…108 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12828669 CAS-Nummer: 2976-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...