1-Ethyl-1H-pyrazole-4-carbaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR1855-1G
OR1855-5G
APOSOR1855-1GEA
29
EUR
APOSOR1855-1G
APOSOR1855-5G
1-Ethyl-1H-pyrazole-4-carbaldehyde 95%
1-Ethyl-1H-pyrazole-4-carbaldehyde
Formel:
C₆H₈N₂O Siedepunkt: 72 °C/1,8mmHg Lagertemperatur: Kühlschrank |
MDL:
MFCD02090888 CAS-Nummer: 304903-10-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...