1H-Benzimidazole-2-carbaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1H-Benzo[d]imidazole-2-carbaldehyde
OR1856-1G
OR1856-250MG
OR1856-5G
APOSOR1856-1GEA
24.8
EUR
APOSOR1856-1G
APOSOR1856-250MG
APOSOR1856-5G
1H-Benzimidazole-2-carbaldehyde 95%
1H-Benzimidazole-2-carbaldehyde
Formel:
C₈H₆N₂O Molecular Weight: 146,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00228045 CAS-Nummer: 3314-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...