5-(Pyridin-3-yl)-1,2,4-oxadiazol-3-amine

Lieferant: Apollo Scientific

Synonyme: 3-Amino-5-(pyridin-3-yl)-1,2,4-oxadiazole

OR18583-1G OR18583-250MG OR18583-500MG
APOSOR18583-1GEA 243 EUR
APOSOR18583-1G APOSOR18583-250MG APOSOR18583-500MG
5-(Pyridin-3-yl)-1,2,4-oxadiazol-3-amine
5-(Pyridin-3-yl)-1,2,4-oxadiazol-3-amine
Formel: C₇H₆N₄O
Molecular Weight: 162,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09864898
CAS-Nummer: 73631-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR