2-Amino-1-(2,5-dimethoxyphenyl)ethanol

Lieferant: Apollo Scientific
Gefahr

Synonyme: alpha-(Aminomethyl)-2,5-dimethoxybenzyl alcohol

OR18586-1G OR18586-100MG OR18586-250MG OR18586-500MG OR18586-2.5G OR18586-5G
APOSOR18586-1GEA 258 EUR
APOSOR18586-1G APOSOR18586-100MG APOSOR18586-250MG APOSOR18586-500MG APOSOR18586-2.5G APOSOR18586-5G
2-Amino-1-(2,5-dimethoxyphenyl)ethanol
2-Amino-1-(2,5-dimethoxyphenyl)ethanol
Formel: C₁₀H₁₅NO₃
Molecular Weight: 197,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03701227
CAS-Nummer: 3600-87-1
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR