5-Brom-m-toluonitril

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-5-methylbenzonitrile

OR18596-5G
APOSOR18596-5GEA 81.2 EUR
APOSOR18596-5G
5-Brom-m-toluonitril
5-Brom-m-toluonitril
Formel: C₈H₆BrN
Molecular Weight: 196,0400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08061970
CAS-Nummer: 124289-21-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR